Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
Investigating Light and Optics
Learn about light and the workings of lenses, mirrors and prisms.
Specifications
For Use With (Application) | Learn about light and the workings of lenses, mirrors and prisms |
---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
S66046
![]() |
Neo/Sci Corporation
201653 |
Investigating Light And Optics Kit | Understand the properties of light, investigate the visible spectrum, explore the reflection of light using mirrors, understand the images formed by converging and diverging lenses, explore polarized light and its uses, study lasers and their uses |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Learn how light travels as well as how it is reflected and refracted
- Investigate with optical instruments such as lenses, mirrors and prisms
- Explore image formation by reflection; light color spectrum formation and dispersion; white, polarized and laser light and their uses in industry and medicine
Activities include:
- Understand the properties of light
- Investigate the visible spectrum
- Explore the reflection of light using mirrors
- Understand the images formed by converging and diverging lenses
- Explore polarized light and its uses
- Study lasers and their uses
Order Info
Class size: 40 students.
Specifications
Learn about light and the workings of lenses, mirrors and prisms |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title