Learn More
InVitroCare™ HTF HEPES Culture Medium
Used for egg retrieval, follicle flushing, ICSI proedures and cryopreservation
Manufacturer:  INVITRO CARE INC 2002
 View more versions of this product
Catalog No. 23-000-901
  Check Availability
Add to cart
Specifications
Description
  • Formulated with EDTA and stabilized form of glutamine
  • HEPES-buffered HTF formulation
  • Does not require use of CO2 incubator
  • Shelf life: 120 days
Description & Specifications
Specifications
Egg retrieval, follicle flushing, ICSI proedures and cryopreservation
EDTA and stabilized form of glutamine
100mL
2°-8°C