InVitroCare™ HTF HEPES Culture Medium

Used for egg retrieval, follicle flushing, ICSI proedures and cryopreservation

Manufacturer: INVITRO CARE INC 20025

 View more versions of this product

Catalog No. 23-000-902

  Check Availability
Add to Cart

    Specifications

    Storage Requirements 2°-8°C
    Quantity 500mL
    For Use With (Application) Egg retrieval, follicle flushing, ICSI proedures and cryopreservation
    Formulation EDTA and stabilized form of glutamine

    • Formulated with EDTA and stabilized form of glutamine
    • HEPES-buffered HTF formulation
    • Does not require use of CO2 incubator
    • Shelf life: 120 days

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit