Learn More
Invitrogen™ Platinum™ SuperFi™ Green PCR Master Mix
Manufacturer:  Invitrogen™ 12359050
 View more versions of this product
Catalog No. 12-359-050
  Check Availability
Add to cart
Specifications
Description
Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase which combines superior fidelity with trusted PlatinumTM hot-start technology for the highest success in PCR. Featuring >100x Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis and other applications benefiting from supreme sequence accuracy.
Description & Specifications
Specifications
  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates 
  • Template generation for sequencing
  • High-throughput PCR
  • Amplification of samples with suboptimal purity
  • Long PCR (up to 20 kb)
  • Fast PCR
Store at -20°C.
Blunt
2X master mix
2X Platinum SuperFi Green PCR Master Mx, 5X SuperFi GC Enhancer, nuclease-free water.
500 rxns