Learn More
Invitrogen™ Platinum™ SuperFi™ Green PCR Master Mix
Manufacturer:  Invitrogen™ 12359250
 View more versions of this product
Catalog No. 12-359-250
  Check Availability
Add to cart
Specifications
Description
Invitrogen™ Platinum™ SuperFi™ DNA Polymerase is a proofreading DNA polymerase which combines superior fidelity with trusted PlatinumTM hot-start technology for the highest success in PCR. Featuring >100x Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis and other applications benefiting from supreme sequence accuracy.