Learn More
AccuPrime™ GC-Rich DNA Polymerase Non-distribution product as customer accommodation.
High-specificity, high-yield PCR amplification of GC-rich templates
Manufacturer:  Invitrogen 12337016
 View more versions of this product
Catalog No. 12-337-016
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Fidelity (vs. Taq): 2X
AccuPrime GC-Rich DNA Polymerase (100 μL, at 2U/μL) is supplied with two 5X reaction buffers and a separate vial (1mL) of 50mMMgSO4. Both buffers contain dNTPs, MgSO4, and AccuPrime accessory proteins. Reaction buffer A (1mL) is for genomic targets. Reaction buffer B (1mL) is for use with cDNA, plasmid, or lambda targets. Sufficient reagents are provided for 200 amplification reactions of 25μL each (at 1 unit of enzyme per reaction).
Store at -20°C.
Gel Compatibility: High
200 reactions