AccuPrime™ Pfx DNA Polymerase Non-distribution product as customer accommodation.
Provides higher fidelity than Pfu DNA polymerase
Manufacturer: Invitrogen 12344032
 View more versions of this product
Catalog No. 12-344-032
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Fidelity (vs. Taq) Fidelity (vs. Taq): 26X
    Gel Compatibility Gel Compatibility: Low
    Includes AccuPrime Pfx DNA Polymerase (500μL, at 2.5U/μL) is provided with 10X AccuPrime reaction mix (5 x 1mL) that contains dNTPs and a separate vial (2 x 1mL) of 50mMMgSO4. Reagents are provided for 1000 amplification reactions of 50μL each.
    Quantity 1000 reactions
    Storage Requirements Store at -20°C.
  • Description & Specifications
Related Products
Product Content Feedback