AccuPrime™ SuperMix I Non-distribution product as customer accommodation.
Efficient PCR amplification, reduce set-up time by half
Manufacturer: Invitrogen 12342028
 View more versions of this product
Catalog No. 12-342-028
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Composition 40mM Tris-HCl (pH 8.4), 100mM KCl, 3mM MgCl2, 400μM dGTP, 400μM dATP, 400μM dTTP, 400μM dCTP, AccuPrime Taq DNA Polymerase, thermostable AccuPrime protein, stabilizers
    Fidelity (vs. Taq) Fidelity (vs. Taq): 2X
    Gel Compatibility Gel Compatibility: Low
    Includes AccuPrime SuperMix I is supplied at 2X concentration to allow approximately 50% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 1000 amplification reactions of 25μL each are provided. AccuPrime SuperMix I Components: 40mM Tris-HCl (pH 8.4), 100mM KCl, 3mM MgCl2, 400μM dGTP, 400μM dATP, 400μM dTTP, 400μM dCTP, AccuPrime Taq DNA Polymerase, thermostable AccuPrime protein, stabilizers. 1000 rxns: 1 x 12.5mL
    Quantity 1000 reactions
    Storage Requirements Store at -20°C in a non-frost-free freezer.
  • Description & Specifications
Related Products
Product Content Feedback