AccuPrime™ SuperMix II Non-distribution product as customer accommodation.

Designed for amplification of genomic DNA templates 200bp-4kb

Manufacturer: Invitrogen 12341020

 View more versions of this product

Catalog No. 12-341-020

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Required Accessories : Template and primers

    Specifications

    Composition 40mM Tris-HCl (pH 8.4); 100mM KCl; 3mM MgCl2, 400μM each dGTP, dATP, dTTP, dCTP; AccuPrime Taq DNA Polymerase; thermostable AccuPrime protein; stabilizers
    Fidelity (vs. Taq) Fidelity (vs. Taq): 2X
    Gel Compatibility Gel Compatibility: Low
    Includes AccuPrime SuperMix II is supplied at 2X concentration to allow approximately 50% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 1000 amplification reactions of 25μL each are provided.AccuPrime SuperMix II Components: 40mM Tris-HCl (pH 8.4); 100mM KCl; 3mM MgCl2, 400μM each dGTP, dATP, dTTP, dCTP; AccuPrime Taq DNA Polymerase; thermostable AccuPrime protein; stabilizers. 1000 rxns: 1 x 12.5mL
    Quantity 1000 reactions
    Storage Requirements Store at -20°C in a non-frost-free freezer.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit