Learn More
Invitrogen™ BioPrime™ Plus Array CGH Genomic Labeling Module Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Enables reproducible labeling of genomic DNA samples for array-based Comparative Genomic Hybridization (CGH)
Manufacturer:  Invitrogen™ 18095014
Catalog No. 18-095-014
Add to cart
Includes: exo-Klenow fragment (40U/µL), 2.5X random primer solution (octamers), control DNA (Salmon Sperm), stop buffer, and distilled water. The BioPrime™ Plus Systems and Modules contain the Alexa Fluor™ modified dNTP mixes or amine modified dNTP mixes and the Alexa Fluor™ NHS Esters. The standard BioPrime™ and BioPrime™ Plus Array CGH Genomic Labeling Systems also contain purification columns and purification buffers.
Description
Description
BioPrime Plus Array CGH Genomic Labeling Module is part of a system that supports:
––High yields of fluorescently labeled genomic DNA
–signal-to-noise ratios
–detection of gene copy number variations.
These systems combine a highly concentrated exo-Klenow fragment of DNA polymerase I and random primers to effectively label genomic targets for sensitive genomic profiling experiments. Exo-Klenow is a mutant of the large fragment of the DNA polymerase I holoenzyme that has both 5' - 3;' and 3' - 5' exonuclease activity removed. The lack of exonuclease activity makes this enzyme ideal for use in random priming protocols, enabling robust yields and efficient incorporation of Alexa Fluor™ AHA fluorescent nucleotides. In the BioPrime Plus Array CGH Genomic Labeling Systems, random octamers anneal to template DNA, providing priming sites for the exo- Klenow enzyme. Alexa Fluor AHA modified nucleotides (direct labeling) or amino-allyl modified nucleotides (indirect labeling) are incorporated as the polymerase extends from the priming sites. The labeled samples are then purified to remove contaminants prior to denaturing and hybridization to a microarray (direct labeling systems) or coupled to the Alexa Fluor NHS ester (indirect labeling systems) prior to hybridization. Available in a direct labeling format.
Array CGH, ChIP-on-Chip, Chromatin Biology, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis, RNAi, Epigenetics and Non-Coding RNA Research
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Shipping Condition: Labeling Module, Dry Ice; Purification Module, Room Temperature
Specifications
Specifications
Module, Genomic Labeling
The BioPrime Core Module is found in all standard BioPrime and BioPrime Plus Systems and Modules. It contains exo-Klenow fragment (40 U/μL), 2.5X random primer solution (octamers), control DNA (Salmon Sperm), stop buffer, and distilled water. The BioPrime Plus Systems and Modules contain the Alexa Fluor modified dNTP mixes or amine modified dNTP mixes and the Alexa Fluor NHS Esters The standard BioPrime and BioPrime Plus Array CGH Genomic Labeling Systems also contain purification columns and purification buffers
Array CGH, ChIP-on-Chip, Chromatin Biology, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis, RNAi, Epigenetics and Non-Coding RNA Research
30 reactions
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.