Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Gateway™ LR Clonase™ II Enzyme mix
Contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps.
Supplier: Invitrogen™ 11791020
Description
- Gateway™ LR Clonase™ II enzyme mix catalyzes the in vitro recombination between an entry clone (containing a gene of interest flanked by attL sites) and a destination vector (containing attR sites) to generate an expression clone
- It contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps
- Guarenteed stable for 6 months when properly stored at -20°C or -80°C
Order Info
Shipping Conditions: Dry ice
Specifications
Gateway™ LR Clonase™ II enzyme mix includes proteinase K solution (2 μg/μL) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored. | |
LR Clonase | |
20 Reactions | |
Clonase™, Gateway™ |
Dry Ice | |
Enzyme Buffer | |
LR Clonase Enzyme Mix |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction