Learn More
Invitrogen™ LR Clonase™ II Plus enzyme
Mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor), and Xis (Excisionase) enzymes.
Supplier: Invitrogen™ 12538120
Includes:
- LR Clonase II Plus enzyme and proteinase K solution (2 μg/μL)
Description
Gateway™ LR Clonase™ enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone. We offer different formats of LR Clonase enzyme mixtures, depending on your application and desired format.
- Gateway™ LR Clonase™ enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes
- They catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate an expression clone
- Different formats of LR Clonase enzyme mixtures are offered depending on the application and desired format
- LR Clonase II Plus enzyme mix is the latest version that offers the highest recombination efficiency available
- It is specifically optimized for both single and multiple fragment cloning
- LR Clonase II Plus enzyme mix is provided in an optimized single mix of enzyme and reaction buffer
- This ensures enzyme stability and ease of use with few pippetting steps for both single and multiple fragment cloning
- Guaranteed stable for 6 months when properly stored at -20°C
Order Info
Shipping Conditions: Dry ice
Specifications
Includes LR Clonase II Plus enzyme and proteinase K solution (2 μg/μL). Store at -20°C. Guaranteed stable for 6 months when properly stored. | |
LR Clonase | |
20 Reactions | |
Clonase™, Gateway™ |
Dry Ice | |
Reaction Buffer | |
LR Clonase Enzyme |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.