Learn More
Cloned AMV First-Strand cDNA Synthesis Kit Non-distribution product as customer accommodation.
Combines the high performance of Invitrogen Cloned AMV RT with the convenience of optimized, prequalified reagents
Manufacturer:  Invitrogen 12328040
 View more versions of this product
Catalog No. 12-328-040
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
First-Strand cDNA Synthesis, RT-PCR
100μL cloned AMV RT (15U/μL), 500μL 5X cDNA synthesis buffer, 2 x 250μL 0.1M DTT, 100μL 10mM dNTP mix, 25μL RnaseOUT (40U/μL), 25μL oligo(dT)20 (50μM), 250μL random hexamers (50ng/μL), 1.25mL DEPC-treated water, product profile sheet
100 reactions (375 units)
113 to 131deg.F
Low
Frozen
Tube
Store at -70deg.C
45 to 55deg.C