Dynabeads™ MyOne™ Carboxylic Acid Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Surface carboxylic acid groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups

Manufacturer: Invitrogen 65011

 View more versions of this product

Catalog No. 65-011

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Applications Hydrophobic/Hydrophilic Protein Purification, Protein and Peptide Purification, Protein Complex Isolation and Analysis, Protein Sample Preparation and Protein Purification, Protein-Protein Interactions, Proteins, Expression, Isolation and Analysis
    Purity Research Grade
    Quantity 2mL
    Sample Type Any Sample Type
    Storage Requirements Store in refrigerator (2 to 8°C).

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit