Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYNAL™ Dynabeads™ M-270 Streptavidin Non-distribution product as customer accommodation.

Gold standard for isolation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets

Manufacturer:  Invitrogen 65305

 View more versions of this product

Catalog No. 65-305


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications Description & Specifications

Specifications

ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), DNA and RNA Purification and Analysis, DNA Extraction, Immunoprecipitation, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis, RNAi, Epigenetics and Non-Coding RNA Research, Sequence-Specific DNA or RNA Purification, Viral RNA/DNA Purification
2.8μm
Research Grade
Any Sample Type
10mg/mL
Dynabeads are supplied in PBS, pH 7.4 with 0.01% Tween™-20 and 0.02% NaN3 added as preservatives
2mL
Store at 2 to 8°C.

Safety and Handling

ShelfLife : 36 months

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit