DYNAL™ Dynabeads™ Pan Mouse IgG, Monoclonal Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Binds monoclonal mouse IgGs of choice for specific cell isolation or depletion

Manufacturer: Invitrogen 11041

 View more versions of this product

Catalog No. 11-041

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Dye Type Dynabeads™ Pan
    Includes 5mL Dynabeads Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is a unique Fc-specific human anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.
    Applications Cell Analysis, Cell Isolation, Cell Isolation and Expansion, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Human Cell Isolation, RNAi, Epigenetics and Non-Coding RNA Research, Secondary Cell Isolation
    No. of Cells 1x107 PBMCs per isolation
    Outputs >95%
    Purification Method Depletion, negative isolation or positive isolation
    Purity Research Grade
    Quantity 5mL
    Sample Type Blood, PBMC, Tissue Digests
    Storage Requirements Storage: 2°C to 8°C.
    Cross Reactivity All species, except mouse

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit