Learn More
DYNAL™ Dynabeads™ Pan Mouse IgG, Monoclonal Non-distribution product as customer accommodation.
Binds monoclonal mouse IgGs of choice for specific cell isolation or depletion
Manufacturer:  Invitrogen 11042
 View more versions of this product
Catalog No. 11-042
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Cell Analysis, Cell Isolation, Cell Isolation and Expansion, ChIP-on-Chip, Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Human Cell Isolation, RNAi, Epigenetics and Non-Coding RNA Research, Secondary Cell Isolation
Dynabeads™ Pan
1x107 PBMCs per isolation
Depletion, negative isolation or positive isolation
5 x 5mL
Storage: 2°C to 8°C.
All species, except mouse
5 x 5mL Dynabeads Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is a unique Fc-specific human anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.
>95%
Research Grade
Blood, PBMC, Tissue Digests