Learn More
Invitrogen™ pBAD-DEST49 Gateway™ Destination Vector Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Designed for rapid cloning with Gateway entry clone using lambda phage site-specific recombination and subsequent high-level, tightly regulated expression in E. coli
Manufacturer:  Invitrogen™ 12283016
Catalog No. 12-283-016
Add to cart
Includes: The pBAD-DEST49 Gateway destination vector includes 6μg of vector.
Description
Description
  • araBAD promoter for tightly regulated expression in E. coli
  • HP-thioredoxin fusion for improved protein yield and solubility
  • Enterokinase cleavage site for efficient cleavage of N-terminal fusion with EKMax™
  • C-terminal V5 and 6xHis tags for efficient detection and purification of fusion proteins
  • R sites for efficient recombination with any attL-flanked Gateway entry vector
Bacterial Expression/Cloning/Gateway Cloning/Protein Expression/Proteins, Expression, Isolation and Analysis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Inducible
araBAD
None
EK (Enterokinase) Recognition Site
His Tag (6x), V5 Epitope Tag, Thioredoxin
pBAD, pDEST
Arabinose
Gateway System Destination Expression Vector
Ampicillin (AmpR)
Gateway™
Gateway™
Safety and Handling
Safety and Handling
SDS
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.