Learn More
Invitrogen™ Gateway™ pDEST™26 Vector Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins
Manufacturer:  Invitrogen™ 11809019
Catalog No. 11-809-019
Add to cart
Includes: All destination vectors are provided lyophilized and supercoiled.
Description
Description
  • Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone
Cloning, Constitutive Expression, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Constitutive
Constitutive Expression
CMV
6μg
His Tag (6X)
Transfection
All destination vectors are provided lyophilized and supercoiled.
Geneticin™ (G-418)
pDEST
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.