Learn More
Invitrogen™ Gateway™ pEF-DEST51 Vector Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or Ultimate™ RF Clone
Manufacturer:  Invitrogen™ 12285011
Catalog No. 12-285-011
Add to cart
Includes: All destination vectors are provided lyophilized and supercoiled.
Requires : Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
Description
Description
Cloning, Constitutive Expression, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Constitutive
EF-1α
Blasticidin
Gateway™
Gateway™
Transfection
Gateway System Destination Expression Vector
Constitutive Expression
His Tag (6x), V5 Epitope Tag
pDEST
Safety and Handling
Safety and Handling
SDS
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.