Learn More
Invitrogen™ Gateway™ pMT-DEST48 Vector Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Designed for rapid cloning with Gateway entry clone using lambda phage site-specific recombination and subsequent expression in Drosophila S2 cells
Manufacturer:  Invitrogen™ 12282018
Catalog No. 12-282-018
Add to cart
Includes: pMT-DEST48 Gateway destination vector is supplied supercoiled and lyophilized.
Description
Description
  • As part of DES Expression System, pMT-DEST48 uses Drosophila metallothionein gene promoter that is induced in S2 cells upon addition of copper sulfate or cadmium chloride to culture medium
  • C-terminal V5 epitope tag for rapid detection with Anti-V5 Antibody
  • C-terminal 6xHis tag for simple purification of recombinant fusion proteins using nickel-chelating resin
  • R sites for efficient recombination with any attL-flanked Gateway entry vector
Cloning, Gateway Cloning, High-level Protein Production in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
MT
DES™, Gateway™
Gateway™
Gateway System Destination Expression Vector
His Tag (6x), V5 Epitope Tag
pMT, pDEST
Safety and Handling
Safety and Handling
SDS
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.