Learn More
Invitrogen™ Gateway™ pT-Rex™-DEST31 Vector Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins
Manufacturer:  Invitrogen™ 12302014
Catalog No. 12-302-014
Add to cart
Includes: All destination vectors are provided lyophilized and supercoiled.
Requires : Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
Description
Description
  • Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone
Cloning, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Inducible
Tetracycline
Gateway System Destination Expression Vector
Regulated Expression
His Tag (6x)
pDEST
Transfection
CMV⁄TO
Geneticin™ (G-418)
Gateway™, T-REx™
Gateway™
Safety and Handling
Safety and Handling
SDS
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.