Lipofectamine™ RNAiMAX Transfection Reagent Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Provides highest transfection efficiencies on widest variety of cell types for siRNA-mediated gene knockdown experiments

Manufacturer: Invitrogen 13778150

 View more versions of this product

Catalog No. 13-778-150

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    For Use With (Application) Synthetic siRNA
    Quantity 1.5mL,1.5mL
    Includes One vial (1.5mL) Lipofectamine RNAiMAX Transfection Reagent
    Storage Requirements Store at 2° to 8°C. Stable for 1 year when stored properly.
    Storage Conditions Store at 2° to 8°C. Stable for 1 year when stored properly.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit