Learn More
Invitrogen™ pBAD TOPO™ TA Expression Kit

Description
Includes
Each pBAD TOPO TA Expression Kit contains two boxes and the LMG194 E. coli stab. The pBAD TOPO TA box contains all of the reagents necessary for PCR (except Taq polymerase), including 200ng of topoisomerase I-activated pBAD-TOPO vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control template and primers, 20% L-arabinose, pBAD primers for sequencing or PCR screening, and an expression control plasmid. The other box contains all of the reagents necessary for transformation, including single-use 50μL aliquots of TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
- Linearized, topoisomerase I-activated vector for 5-minute cloning of Taqamplified PCR products
- araBAD promoter for tightly regulated expression in E. coli V5 epitope tag for detection with Anti-V5 Antibody
- C-terminal polyhistidine (6xHis) tag for purification using nickel-chelating resin and detection with Anti-His(C-term) Antibody
- Enterokinase cleavage site for removal of N-terminal leader peptide
Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

Specifications
Specifications
Constitutive or Inducible System | Inducible |
Inducing Agent | Arabinose |
Promoter | araBAD |
Product Type | TA Expression Kit |
Selection Agent (Eukaryotic) | None |
Content And Storage | Each pBAD TOPO™ TA Expression Kit contains two boxes and the LMG194 E. coli stab. The pBAD TOPO™ TA box contains all of the reagents necessary for PCR (except Taq polymerase), including 200 ng of topoisomerase I-activated pBAD-TOPO™ vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control template and primers, 20% L-arabinose, pBAD primers for sequencing or PCR screening, and an expression control plasmid. Store at -20°C. The One Shot™ box contains all of the reagents necessary for transformation, including single-use 50 μl aliquots of One Shot™ TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Store One Shot™ cells at -80°C. Store the LMG194 E. coli stab at at 2 - 8°C. All reagents are guaranteed stable for 6 months when properly stored. |
Antibiotic Resistance Bacterial | Ampicillin (AmpR) |
Bacterial or Yeast Strain | LMG194, TOP10 |
Cleavage | EK (Enterokinase) Recognition Site |
Expression System | E. coli |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.