Learn More
Platinum™ GenoType Tsp DNA Polymerase Non-distribution product as customer accommodation.
Built-in hot start allows for room temperature reaction
Manufacturer:  Invitrogen 11448024
 View more versions of this product
Catalog No. 11-448-024
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Fidelity (vs. Taq): 1X
Platinum GenoType TspDNA Polymerase is supplied with 10X PCR Buffer (200mM Tris-HCl (pH 8.4), 500mM KCl), and a vial of 50mM MgCl2.
Store at -20°C. Guaranteed stable for 6 months when properly stored.
Gel Compatibility: Low
250 units