Learn More
Platinum™ Pfx DNA Polymerase Non-distribution product as customer accommodation.
Built-in hot start
Manufacturer:  Invitrogen 11708013
 View more versions of this product
Catalog No. 11-708-013
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Fidelity (vs. Taq): 26X
Platinum Pfx DNA Polymerase (100 units total, at 2.5U/μL) is a supplied with a vial (1mL) of 10X PCRx Enhancer solution, a vial (1mL) of 10X Pfx amplification buffer, and a vial (1mL) of 50mM MgSO4. Designed for 100 reactions (at 1 unit each in a 50μL reaction volume).
Store at -20°C.
Gel Compatibility: Medium
100 reactions