Platinum™ PCR SuperMix Non-distribution product as customer accommodation.

Ready-to-use mixture reduces set up time by 50%

Manufacturer: Invitrogen 11306016

 View more versions of this product

Catalog No. 11-306-016

  N/A
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Required Accessories : Template and primers
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    For Use With (Application) Hot Start PCR, PCR, PCR and Real-Time PCR, Routine PCR
    Includes Platinum PCR SuperMix is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 5000 amplification reactions of 50μL each are provided. Platinum PCR SuperMix Components: 22U/mL complexed recombinant Taq DNA polymerase with Platinum Taq Antibody, 22mM Tris-HCl (pH 8.4), 55mM KCl, 1.65mM MgCl2, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP, and stabilizers. 100 rxns: 4 x 1.125mL
    No. of Reactions 100.0
    Overhang 3'-A
    Quantity 100 reactions
    Size (Final Product) 5kb or less

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit