Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG w/ROX
qPCR SuperMixes with ROX for ABI instruments are ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.
Supplier: Invitrogen™ 11743100
Includes: Platinum™ Quantitative PCR SuperMix-UDG with ROX; Magnesium Chloride (50mM)
Description
qPCR SuperMixes with ROX are available in two formats:- Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dye
- Platinum™ qPCR SuperMix-UDG with ROX offers superior performance with either fluorogenic primers (e.g., LUX™ Primers) or probe-based detection systems (e.g., TaqMan™)
Order Info
Guaranteed stable for 6 months when properly stored. Shipping Conditions: Dry Ice.
Specifications
• 2 × 1.25 ml Platinum™ Quantitative PCR SuperMix-UDG with ROX • 1 ml Magnesium Chloride (50 mM) Store at -20°C in the dark. |
|
High | |
Platinum Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System | |
Quantitative PCR SuperMix-UDG | |
100 Reactions | |
2X | |
100 Reactions | |
2.5 mL |
Primer-probe | |
qPCR | |
Standard | |
Platinum™ | |
Dry Ice | |
Gene Expression | |
2 X | |
DNA (Genomic), cDNA |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction