Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG
Is a ready-to-use mixture for high-specificity, real-time quantitative DNA amplification, containing UDG to prevent carryover contamination (1,2).
Supplier: Invitrogen™ 11730025
Includes: 2X concentration [40mM Tris-HCl (pH 8.4), 100mM KCl, 6mM MgCl2 , 400μM dATP, 400μM dCTP, 400μM dGTP, 800μM dUTP, 40 units/mL UDG, 60 units/ml Platinum™ Taq DNA Polymerase, stabilizers], vial of 50mM MgCl2
Description
- Highly sensitive and specific
- Allows room temperature reaction assembly
- Easy to use: just add template and fluorogenic primers or probes for detection
- Includes a vial of 50mM MgCl2 to accommodate a variety of primer/template PCR systems
- Compatible with most real-time instruments
Order Info
Guaranteed stable for 6 months when properly stored. Guaranteed minimum shelf life is 60 days. Shipping Conditions: Dry Ice.
Specifications
Platinum™ Quantitative PCR SuperMix-UDG is supplied in a 2X concentration [40 mM Tris-HCl (pH 8.4), 100 mM KCl, 6 mM MgCl2 , 400 μM dATP, 400 μM dCTP, 400 μM dGTP, 800 μM dUTP, 40 units/ml UDG, 60 units/ml Platinum™ Taq DNA Polymerase, stabilizers]. A vial of 50 mM MgCl2 is also provided. Store at -20°C. Guaranteed stable for 6 months when properly stored. | |
High | |
Standard | |
Platinum™ | |
Dry Ice | |
Gene Expression | |
DNA (Genomic), cDNA |
Primer-probe | |
qPCR | |
7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ | |
Quantitative PCR SuperMix-UDG | |
500 Reactions | |
500 Reactions |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction