Learn More
Invitrogen™ Platinum™ SYBR™ Green qPCR SuperMix-UDG w/ROX
Description
Includes
2X SuperMix with SYBR Green I and ROX Reference Dyes, and 50mM MgCl2
Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. Uracil-DNA Glycosylase (UDG) prevents carryover contamination in conjunction with dUTP (included) for improved specificity. ROX reference dye is included in the mix for maximum convenience. The ROX reference dye premixed at optimal concentration for ABI instruments.
Order Info
Shipping Conditions: Dry Ice
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | SYBR |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System |
| Product Line | Platinum™, SYBR™ |
| Product Type | Real Time PCR SYBR Master Mix |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.