Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ SYBR™ Green qPCR SuperMix-UDG Non-distribution product as customer accommodation.

Provides a sensitive and convenient system for real-time PCR detection with SYBR™ Green I dye.

Manufacturer:  Invitrogen™ 11733046

 View more versions of this product

Catalog No. 11-733-046


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

The SuperMix format combines amplification and detection reagents into a ready-to-go mix to make set-up easy and ensure reproducible results.

  • Platinum™ Taq DNA Polymerase for sensitive amplification from as few as 10 copies of target
  • 100% dUTP and UDG enzyme for superior control of carryover contamination, saving time by reducing the number of failed experiments
  • Separate tubes of ROX reference dye and BSA for easy optimization on 96-well plates and glass capillary tube instruments
  • SYBR™ Green I dye for simple and easy detection of real-time product

Order Info

Shipping Conditions: Dry Ice

Description & Specifications

Specifications

Chromatin biology, real time PCR (qPCR), real time PCR-based gene expression profiling
500 Reactions
Store all components at -20°C.
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, BioRad iCycler iQ, Cepheid SmartCycler, GeneAmp™ 5700, StepOne™, StepOnePlus™, ViiA™ 7 System
Standard
Platinum™ SYBR™ Green qPCR SuperMix-UDG (12.5mL); 50mM Magnesium Chloride (2 x 1mL); ROX Reference Dye (500μL); 20X BSA (1.3mL). Sufficient reagents are provided for 100 reactions of 50μL each.
Sufficient reagents are provided for 500 reactions of 50μL each
Detection Method: SYBR; PCR Method: qPCR
DNA (Genomic), cDNA
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit