Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYBR GreenER™ qPCR SuperMix Universal Non-distribution product as customer accommodation.

Incorporate the latest high-performance technology to provide the most reliable gene expression data on a variety of platforms including ABI 7500, Corbett Rotor-Gene™ etc.

Manufacturer:  Invitrogen™ 1176202K

 View more versions of this product

Catalog No. 11-762-02K


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

SYBR™ GreenER™ qPCR SuperMixes Universal incorporate the latest high-performance technology to provide the most reliable gene expression data on a variety of platforms, including ABI 7500, Corbett Rotor-Gene™, MJ Chromo4™ and Opticon™, and Stratagene instruments. The SYBR™ GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility with. A novel dsDNA-binding dye exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR™ Green I, while using the same instrument filters and settings. An optimized buffer system improves sensitivity and provides excellent long-term stability. Uracil DNA glycosylase reduces carryover contamination in qPCR.
Eight cDNA targets and eight genomic DNA targets were amplified from 1ng of cDNA and 6pg of gDNA, respectively, on the ABI PRISM™ 7900HT Sequence Detection System using the SYBR™ GreenER™ qPCR SuperMix for ABI PRISM™ instruments. For all 16 targets, detection was positive in the presence of template but negative for all no-template controls.

  • The SYBR™ GreenER™ qPCR SuperMix kits include a new dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range

Consistent Specificity and Low-Copy Detection:
  • The SYBR™ GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets
  • The reliability of the SYBR™ GreenER™ qPCR reagent system also translates to greater sensitivity
  • With SYBR™ GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA
Description & Specifications

Specifications

Chromatin biology, real time PCR (qPCR), real time PCR-based gene expression profiling
2000 Reactions
Detection Method: SYBR; PCR Method: qPCR
DNA (Genomic), cDNA
2X SYBR™ GreenER™ qPCR SuperMix and a separate tube of ROX reference dye.
Store at 4°C upon receipt.
7500 System, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
Standard

Safety and Handling

ShelfLife : Guaranteed stable for 6 months when properly stored.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit