Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ III Platinum™ SYBR™ Green One-Step qPCR Kit With ROX Non-distribution product as customer accommodation.

Combine a powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.

Manufacturer:  Invitrogen™ 11746100

 View more versions of this product

Catalog No. 11-746-100


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient, high-throughput one-step format. SuperScript™ III Reverse Transcriptase produces higher yields of full-length cDNA for broader gene representation. Platinum™ Taq DNA Polymerase activates faster and more completely than other hot-start enzymes to deliver more sensitive amplification. ROX reference dye premixed at optimal concentration for ABI instruments.

  • Provides a highly efficient, cost-effective qRT-PCR system with SYBR Green I detection

Order Info

Shipping Conditions: Dry Ice

Description & Specifications

Specifications

Real time PCR (qPCR), real time PCR-based gene expression profiling
100 Reactions
Detection Method: SYBR; PCR Method: 1-Step qRT-PCR
RNA
100 Reactions
SuperScript™ III RT/Platinum™ Taq Mix, 2X Reaction Mix (containing SYBR™ Green I, ROX Reference Dye, dNTPs, and MgSO4), and 50mM MgSO4. SuperScript™ III Platinum™ One-Step qRT-PCR Kit with ROX includes SuperScript™ III RT/Platinum™ Taq Mix, 2X Reaction Mix (containing ROX Reference Dye, dNTPs and MgSO4), and 50mM MgSO4.
Store all components at -20°C. Store ROX Reference dye in the dark. Store in freezer (-5° to -30°C).
7000 System, 7300 System, 7700 System, 7900HT System
Standard

Safety and Handling

ShelfLife : Guaranteed stable for 6 months when properly stored.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit