Learn More
Invitrogen™ Quant-iT™ RiboGreen™ RNA Reagent Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
One of most sensitive fluorescent dyes available for quantitation of RNA in solution
Manufacturer:  Invitrogen™ R11491
Catalog No. R11491
Add to cart
Description
Description
  • Used with linear fluorescence detection in range of 1-200ng of RNA
  • Available in kit complete with buffer and RNA standard (Cat. No. O11490)
DNA and RNA Purification and Analysis, Nucleic Acid Quantitation, RNA Quantitation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
Microplate Reader
Store in refrigerator (2–8°C) and protect from light.
Room Temperature
RNA
1-200 ng
Quant-iT™, RIBOGREEN®
SDS
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.