Learn More
Invitrogen™ SuperScript™ Plasmid System for cDNA Synthesis and Plasmid Cloning with Gateway™ Technology Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Designed for synthesis of double-stranded cDNA from purified mRNA population
Manufacturer:  Invitrogen™ 18248013
Catalog No. 18-248-013
Add to cart
Includes: The SuperScript Plasmid System with Gateway Technology includes components listed above.
Description
Description
The SuperScriptPlasmid System is designed for synthesis of double-stranded cDNA from a purified mRNA population. The cDNA is suitable for directional cloning into the pSPORT1 vector or the pCMV.
  • SPORT6 plasmid vector for subsequent generation of cDNA libraries and use of GatewayTechnology
  • Results in greater cDNA yields and increased full-length cDNAs with SuperScriptII RT
  • Offers simplified primer-adapter strategy for directional cloning
  • Offers one-tube format for first- and second-strand reactions, improving cDNA yields
  • Fractionates cDNA using efficient and convenient pre-packed columns
  • Ligates inserts into a pre-cut Not I-Sal I vector prepared to minimize background
Cloning, cDNA Libraries and Library Construction
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
3 reactions
Store all cDNA cloning and ligation reagents at -20°C. Store cDNA size fractionation columns at 4°C.
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.