Learn More
Invitrogen™ Terminal Deoxynucleotidyl Transferase, recombinant
Performance and quality tested
Supplier: Invitrogen™ 10533065
Alert:
Hazard Warning: Toxic; potassium cacodylate contained in reaction buffer. Also contains cobalt chloride, a highly toxic chemical. See MSDS.
Includes: Vial of 5X buffer [500mM potassium cacodylate (pH 7.2), 10mM CoCl2 , 1mM DTT]
Description
Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is DNA polymerase that catalyzes addition of deoxynucleotides to 3fi hydroxyl terminus of DNA.
- Source: Purified from baculovirus clone of calf thymus TdT
- Performance and Quality Testing: Endonuclease, 3' and 5' exodeoxyribonuclease, and levels of incorporation tested
- Unit Definition: One unit incorporates 1nmol dATP into acid-precipitable material in 1 hr. at 37°C, using d(pA)50 as primer
- Unit Reaction Conditions: 0.2M potassium cacodylate (pH 7.2), 10mM MgO4 C4 H6 , 1mM 2-mercaptoethanol, 0.5mg/mL BSA, 100 flM d(pA)50, 1mM [3H]dATP, and enzyme in 0.15mL for 1 h at 37°C
Cloning; Restriction Enzyme Cloning; Homopolymer tailing of vector and insert for cloning; Labeling oligonucleotides with biotin, 32P- or 35S-label; Apoptosis (TUNEL)
Order Info
Shipping Conditions: Approved for shipment on wet or dry ice
Specifications
Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is supplied with vial of 5X buffer [500 mM potassium cacodylate (pH 7.2), 10 mM CoCl2 , 1 mM DTT]. Store at -20°C. | |
Terminal Deoxynucleotidyl Transferase | |
500 U |
Approved for shipment on Wet or Dry Ice | |
5X Buffer, Reaction Buffer | |
TdT |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.