ThermoScript™ Reverse Transcriptase Non-distribution product as customer accommodation.

An avian RT with reduced RNase H activity designed for RT-PCR

Manufacturer: Invitrogen 12236014

 View more versions of this product

Catalog No. 12-236-014

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.


    Specifications

    Format Frozen,Frozen
    Temperature (English) Operating 149°F
    Temperature (Metric) Operating 65°C
    Test Sensitivity Medium
    Packaging Type Tube
    Includes 5X cDNA synthesis buffer (250mM Tris acetate (pH 8.4), 375mM potassium acetate, 40mM magnesium acetate, and stabilizers), and 100mM DTT
    Concentration 15U/μL,15U/μL
    Quantity 25 reactions (375 units),25 reactions (375 units)
    Storage Requirements Store at -70deg.C

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit