ThermoScript™ RT-PCR System for First-Strand cDNA Synthesis Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Optimized for generating first-strand cDNA from total or poly(A)+ RNA

Manufacturer: Invitrogen 11146016

 View more versions of this product

Catalog No. 11-146-016

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Includes 100μL ThermoScript RT (15U/μL); 500μL 5X cDNA Synthesis Buffer; 250μL 0.1M DTT; 2 × 250μL 10mM dNTP Mix; 100μL RNaseOUT (40U/μL); 100μL Oligo (dT)20 (50μM); 250μL Random Hexamers (50ng/μL); 1.25mL DEPC-Treated Water; 2 × 50μL E. coli Rnase H (2U/μL)
    Temperature (English) Operating 122° to 149°F
    Temperature (Metric) Operating 50° to 65°C
    For Use With (Application) First-Strand cDNA Synthesis, RT-PCR
    Primer Random Primers, Oligo dT Primers
    Quantity 100 reactions,100 reactions
    Storage Requirements Store at -20deg.C

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit