Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)
UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v) is used in the purification of nucleic acids. This reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl.
Supplier: Invitrogen™ 15593049
Description
- Reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl
- When mixtures are extracted with Phenol:Chloroform:Isoamyl Alcohol, proteins are denatured and collected in the organic phase or at the interphase, while nucleic acids remain in the aqueous phase
- UltraPure™ Phenol:Chloroform:lsoamyl Alcohol contains no preservatives
- It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles
- The appearance of the solution is evaluated at room temperature
- No RNase or DNase activity is detected
- Store in the refrigerator at 2°C to 8°C
Order Info
Shipping Condition: Room temperature
Specifications
Phenols | |
8.05 | |
Molecular Biology Grade | |
Room Temperature |
Store in refrigerator (2–8°C). | |
UltraPure™ | |
400 mL | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction