Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Involucrin Ab-1, Mouse Monoclonal Antibody

Thermo Scientific™ Involucrin Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$470.00 - $679.50

Specifications

Antigen Involucrin Ab-1
Clone SY5
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS126P
View Documents
Thermo Scientific™
MS126P
Purified Ab with BSA and Azide 1mL
Each for $679.50
Add to cart
 
MS126P1
View Documents
Thermo Scientific™
MS126P1
Purified Ab with BSA and Azide 500μL
Each for $537.50
Add to cart
 
MS126P1ABX
View Documents
Thermo Scientific™
MS126P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $470.00
Add to cart
 
MS126PABX
View Documents
Thermo Scientific™
MS126PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $601.00
Add to cart
 
MS126B
View Documents
Thermo Scientific™
MS126B
Biotinylated Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS126P0
View Documents
Thermo Scientific™
MS126P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS126R1
View Documents
Thermo Scientific™
MS126R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS126B0
View Documents
Thermo Scientific™
MS126B0
Biotinylated Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS126B1
View Documents
Thermo Scientific™
MS126B1
Biotinylated Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS126R7
View Documents
Thermo Scientific™
MS126R7
Ready-to-use for IHC (UVLP) 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Thermo Scientific™ Involucrin Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results. 

Involucrin is expressed in a range of stratified squamous epithelia, including the cornea. In normal epidermis, it is first expressed in the upper spinous layers, and in keratinocyte cultures it is expressed by all cells that have left the basal layer. Involucrin expression is abnormal in squamous cell carcinomas and premalignant lesions, and is reduced in severe dysplasias of the larynx and cervix.

Host Species: Mouse

Clone: SY5

Isotype: IgG1

Species Reactivity: Human, Gorilla, Owl, Monkey, Pig, Dog and Rat. Others not known.

Epitope: Codon 421-568

Immunogen: Human keratinocytes' involucrin

Molecular Weight: 66-170kDa

Positive Control: LS174T cells, Skin

Cellular Localization: Cytoplasmic Ab-1 stains the involucrin in a variety of sizes: 170kDa in MCF-7 cells, a doublet of ~115kDa and 150kDa in gorilla and owl monkey, 66kDa in dog, and a doublet of 105kDa in pig.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Involucrin Ab-1
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Cytoskeletal Protein
Human keratinocytes involucrin
SY5
Monoclonal
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit