Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Involucrin Mouse anti-Human, Porcine, Canine, Primate, Monkey, Clone: rIVRN/827, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30755920UG
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate, Monkey samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Involucrin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 0.1-0.2 ug/ml | |
involucrin | |
Recombinant human full-length Involucrin protein (IVRN) | |
20 μg | |
Extracellular Matrix | |
3713 | |
Store at 4°C. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rIVRN/827 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Porcine, Canine, Primate, Monkey | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction