Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Involucrin Rabbit anti-Human, Porcine, Canine, Primate, Monkey, Clone: IVRN/2113R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30757720UG
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate, Monkey samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Involucrin | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| involucrin | |
| Recombinant human full-length Involucrin protein (IVRN) | |
| 20 μg | |
| Extracellular Matrix | |
| 3713 | |
| Store at 4°C. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| IVRN/2113R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Pig, Canine, Primate, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction