Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, 0.1 N standard solution, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
7553-56-2,7681-11-0,7732-18-5
I2
253.809
MFCD00011355
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode
807
CHEBI:17606
molecular iodine
II
7553-56-2,7681-11-0,7732-18-5
I2
253.809
MFCD00011355
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode
807
CHEBI:17606
molecular iodine
II
Volumetric standard solution

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Linear Formula I2
Fieser 01,495; 02,200; 03,159; 04,258; 05,346; 06,293; 07,179; 08,256; 09,248; 10,210; 11,261; 12,235; 13,133; 14,181; 15,172; 16,182; 17,153
Merck Index 15, 5057
Formula Weight 253.81
Physical Form Liquid
Concentration 0.0998 to 0.1002N (20°C)
Density 1.0200g/mL
Packaging Glass bottle
Color Brown
Quantity 2.5L
Chemical Name or Material Iodine, 0.1N Standard Solution
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 231-442-4

RTECSNumber : NN1575000

TSCA : TSCA