Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine, 1.0N Standardized Solution, Alfa Aesar™
Click to view available options
Quantity:
500mL
1L
Specifications
Solubility Information | Miscible with chloroform,carbon tetrachloride,carbon disulfide,cyclohexane,methanol,ethyl acetate,toluene,benzene,n-hexane,butan-2-ol,bromoethane,n-heptane,glycerol and diethyl ether. |
Formula Weight | 253.81 |
Physical Form | Liquid |
Quantity | 1L |
Chemical Name or Material | Iodine |
Safety and Handling
P260-P264b-P270-P314-P501c
H335-H372
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 231-442-4
TSCA : Yes
Recommended Storage : Ambient temperatures