Iodine, 1.0N Standardized Solution, Alfa Aesar™

Manufacturer: Alfa Aesar 35632AP

Quantity 500mL

 View more versions of this product

Catalog No. AA35632AP

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula I2
    Formula Weight 253.81
    MDL Number MFCD00011355
    Chemical Name or Material Iodine

    Specifications

    Formula Weight 253.81
    Quantity 500mL
    Physical Form Liquid
    Molecular Formula I2
    MDL Number MFCD00011355
    Chemical Name or Material Iodine

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit