Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Acros Organics™

Click to view available options
Quantity:
100g
500g
7553-56-2
I2
253.809
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode
807
CHEBI:17606
molecular iodine
II
7553-56-2
I2
253.809
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode
807
CHEBI:17606
molecular iodine
II

C-X bond formation reagent

Linear Formula I2
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in aq.sol. of hi or iodines, soluble in alcohol, carbon disulfide, chloroform,, ether, ccl4 and glycerol
Formula Weight 253.81
Percent Purity 99.5 to 100.5% (Iodometry)
Grade Analysis
Calcium (Ca) 10ppm max.
Chloride and Bromide 0.005% max. (as Cl)
Copper (Cu) 5ppm max.
Insoluble Matter Passes test
Iron (Fe) 50ppm max.
Lead (Pb) 5ppm max.
Magnesium (Mg) 5ppm max.
Packaging Glass bottle
Potassium (K) 50ppm max.
Sodium (Na) 50ppm max.
Viscosity 2.27 mPa.s (116°C)
Zinc (Zn) 5ppm max.
Boiling Point 185.0°C
Melting Point 113.0°C
Quantity 100g
Chemical Name or Material Iodine
Show More Show Less

GHS H Statement Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. May be corrosive to metals.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid release to the environment. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger