Iodine-Bromine Solution, Hanus, for Iodine Absorption Number of Fats and Oils, Ricca Chemical

For Analysis of Iodine Absorption Number of Fats and Oils

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 410032

Packaging Amber Glass
Quantity 1L

 View more versions of this product

Catalog No. 4100-32

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64-19-7,7553-56-2,7726-95-6
    CAS Min % 96.0,1.21,0.85
    CAS Max % 100.0,1.26,0.89
    Physical Form Liquid
    Chemical Name or Material Iodine-Bromine Solution, Hanus

    Specifications

    Color Dark brown
    Physical Form Liquid
    Quantity 1L
    CAS Max % 0.89,100.0,1.26
    CAS Min % 96.0,1.21,0.85
    Packaging Amber Glass
    CAS 7726-95-6,64-19-7,7553-56-2
    Chemical Name or Material Iodine-Bromine Solution, Hanus

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit