Iodine, D'Antoni, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 41034

Packaging Glass amber
Quantity 120mL

 View more versions of this product

Catalog No. 41034

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,7681-11-0,7553-56-2
    CAS Min % 96.0,0.96,0.96
    CAS Max % 100.0,1.0,1.0
    Physical Form Liquid
    Chemical Name or Material Iodine, D'Antoni

    Specifications

    Physical Form Liquid
    Color Dark brown
    Quantity 120mL
    CAS Max % 1.0,100.0,1.0
    CAS Min % 96.0,0.96,0.96
    CAS 7681-11-0,7732-18-5,7553-56-2
    Packaging Glass amber
    Chemical Name or Material Iodine, D'Antoni
  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit