Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine (Iodine-Iodide), 0.01N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
Packaging:
Narrow Mouth Amber Glass Bottle
7732-18-5
I
7732-18-5
I

High quality chemicals for laboratory and specialized industrial use

Formula Weight 126.9
Packaging Narrow Mouth Amber Glass Bottle
Quantity 1L
Specific Gravity 1L = 1.00kg
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Iodine (Iodine-Iodide), 0.01N Volumetric Solution