Iodine (Iodine-Iodide), 0.100 Normal, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner R40000004X4C

Packaging Amber glass
Quantity 16L

 View more versions of this product

Catalog No. 40-001-CS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7553-56-2,7681-11-0
    Physical Form Liquid
    Assay Percent Range 0.1000 ± 0.0002 N at 20°C
    Chemical Name or Material Iodine (Iodine-Iodide), 0.100 N (N/10)

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 16L
    Solvent or Matrix Nonaqueous Solution
    Assay Percent Range 0.1000 ± 0.0002 N at 20°C
    CAS 7553-56-2,7681-11-0
    Odor Characteristic iodine odor
    Packaging Amber glass
    Chemical Name or Material Iodine (Iodine-Iodide), 0.100 N (N/10)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit